missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO-1 Antibody (A180), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 39 publications
£139.00 - £436.00
Specifications
| Antigen | NQO-1 |
|---|---|
| Clone | A180 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1-2 ug/mL, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Flow (Intracellular) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18706663
|
Novus Biologicals
NB200-209SS |
0.025 mL |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18709693
|
Novus Biologicals
NB200-209 |
0.1 mL |
£436.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NQO-1 Monoclonal specifically detects NQO-1 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
| NQO-1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Lipid and Metabolism, Signal Transduction | |
| P15559 | |
| 1728 | |
| Full length recombinant NQO1 from human lung [UniProt# P15559] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 31 kDa |
| A180 | |
| Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1-2 ug/mL, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Flow (Intracellular) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Canine, Primate | |
| Azoreductase, DHQU, DIA4azoreductase, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), dioxin-inducible 1, DTDEC 1.6.5.2, DT-diaphorase, Menadione reductase, NAD(P)H dehydrogenase [quinone] 1, NAD(P)H dehydrogenase, quinone 1, NAD(P)H:menadione oxidoreductase 1, NAD(P)H:Quinone acceptor oxidoreductase type 1, NAD(P)H:quinone oxidoreductase 1, NAD(P)H:quinone oxireductase, NMOR1diaphorase-4, Phylloquinone reductase, QR1NMORI, Quinone reductase 1 | |
| NQO1 | |
| IgG1 | |
| Protein G purified | |
| Does not cross-react with NQO2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title