missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO-1 Antibody (A180), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 39 publications
Brand: Novus Biologicals NB200-209
This item is not returnable.
View return policy
Description
NQO-1 Monoclonal specifically detects NQO-1 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications
| NQO-1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1-2 ug/mL, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Flow (Intracellular) | |
| Azoreductase, DHQU, DIA4azoreductase, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), dioxin-inducible 1, DTDEC 1.6.5.2, DT-diaphorase, Menadione reductase, NAD(P)H dehydrogenase [quinone] 1, NAD(P)H dehydrogenase, quinone 1, NAD(P)H:menadione oxidoreductase 1, NAD(P)H:Quinone acceptor oxidoreductase type 1, NAD(P)H:quinone oxidoreductase 1, NAD(P)H:quinone oxireductase, NMOR1diaphorase-4, Phylloquinone reductase, QR1NMORI, Quinone reductase 1 | |
| Mouse | |
| 31 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Lipid and Metabolism, Signal Transduction | |
| 1728 | |
| Human, Mouse, Rat, Canine, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| A180 | |
| Unconjugated | |
| P15559 | |
| NQO1 | |
| Full length recombinant NQO1 from human lung [UniProt# P15559] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Does not cross-react with NQO2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction