missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOX4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14347-1-AP-150UL
This item is not returnable.
View return policy
Description
Oxygen sensing is essential for homeostasis in all aerobic organisms. A phagocyte-type oxidase, similar to that responsible for the production of large amounts of reactive oxygen species (ROS) in neutrophil granulocytes, with resultant antimicrobial activity, has been postulated to function in the kidney as an oxygen sensor that regulates the synthesis of erytheropoietin in the renal cortex. NOX4 has a role as a redox messenger in the activation of intracellular signaling pathways leading (or contributing) to mitochondriogenesis, cell survival, and differentiation in hematopoietic stem cells. Data suggests that NOX4 provides a novel link between the insulin receptor and the generation of cellular reactive oxygen species that enhances insulin signal transduction.Specifications
| NOX4 | |
| Polyclonal | |
| Unconjugated | |
| NOX4 | |
| Kidney oxidase 1, KOX, KOX 1, NADPH oxidase 4, NOX4, Renal NAD(P)H oxidase, RENOX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 50490, 50507 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JHI8, Q9NPH5 | |
| Nox4 | |
| NOX4 Fusion Protein Ag5687 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction