missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Chromata ChIP Histone H3 [Monomethyl Lys9] Kit (ChIP)

Product Code. 18267311 Shop All Bio Techne Products
Click to view available options
Quantity:
25 Immunoprecipitations
Unit Size:
25 reactions
This item is not returnable. View return policy

Product Code. 18267311

Brand: Novus Biologicals™ NBP171713

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair

Applications: Chromatin Immunoprecipitation

Kit contains all the necessary components to perform 25 Immunoprecipitation experiments including a positive control antibody. This ChIP kit is specific for Histone H3 monomethyl Lysine 9.

TRUSTED_SUSTAINABILITY

Specifications

Kit Contents 10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair
Content And Storage Storage of components varies. See protocol for specific instructions.
Quantity 25 Immunoprecipitations
For Use With (Application) Chromatin Immunoprecipitation of DNA from fixed Hela cells alongside a no antibody (No Ab) control
Target Chromata ChIP Histone H3
Type ChIP Histone H3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.