Learn More
Invitrogen™ NovaFluor™ Antibody Conjugation Kits
Description
NovaFluor Conjugation Kits provide a simple workflow to conjugate a blue, yellow-green or red excitable fluorophore to your antibody of choice in just 3 easy steps. The fluorescence-by-design Phiton platform enables the addition of parameters to your current panels in discovery research and resolves stained populations with near-zero cross excitation, with a lack of emission in the violet channels that plague other fluorophores.
NovaFluor Conjugation Kit provides features including:
- A clean and bright antibody label to increase the number of parameters on conventional and spectral flow cytometers
- Resolve populations with limited/near-zero cross excitation and the lowest spread performance of any blue, yellow-green or red excited fluorescent labels
- Simple 3-step protocol, with 60 sec. of hands-on time for conjugating 10–100 μg of antibody per label
- Up to 70% less excitation when compared to PE tandems, resulting in decreased spillover and higher resolution
3-step conjugation method
- Attach NovaFluor Linker to antibody of choice
- Remove excess NovaFluor Linker from DNA-conjugate antibody
- Anneal NovaFluor to oligo-conjugated antibody
Specifications
Specifications
| For Use With (Application) | Flow Cytometry |
| Quantity | 100 μg |
| Product Type | Conjugation Kit |
| Label or Dye | NovaFluor™ Yellow 610 |
| Product Line | NovaFluor |
| Emission | 610 nm |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.