missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOV/CCN3 Rabbit anti-Human, Clone: 130, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £448.00
Specifications
| Antigen | NOV/CCN3 |
|---|---|
| Clone | 130 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274926
|
Novus Biologicals
NBP2-89369-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18247057
|
Novus Biologicals
NBP2-89369-100UL |
100 μg |
£448.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOV/CCN3 Monoclonal specifically detects NOV/CCN3 in Human samples. It is validated for ELISA.Specifications
| NOV/CCN3 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 4856 | |
| IgG | |
| Protein A purified |
| 130 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CCN family member 3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
| Recombinant Human NOV/CCN3 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title