missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOV/CCN3 Rabbit anti-Human, Clone: 130, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89369-50UL
This item is not returnable.
View return policy
Description
NOV/CCN3 Monoclonal specifically detects NOV/CCN3 in Human samples. It is validated for ELISA.
Specifications
| NOV/CCN3 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 4856 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 130 | |
| ELISA 1:5000-1:10000 | |
| CCN family member 3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
| Recombinant Human NOV/CCN3 Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction