missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin, Thermo Scientific™

A synthetic fluoroquinolone antibiotic

£24.60 - £151.00

Chemical Identifiers

CAS 70458-96-7
Molecular Formula C16H18FN3O3
Molecular Weight (g/mol) 319.336
MDL Number MFCD00079532
InChI Key OGJPXUAPXNRGGI-UHFFFAOYSA-N
Synonym Baccidal; Sebercim
PubChem CID 4539
ChEBI CHEBI:100246
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15464979
SDS View Documents
Thermo Scientific Alfa Aesar
J62652.03
1g
ONLINE EXCLUSIVE PRICE
£25.10
£24.60 / 1g
Save £0.50
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15474979
SDS View Documents
Thermo Scientific Alfa Aesar
J62652.06
5g
ONLINE EXCLUSIVE PRICE
£53.60
£52.40 / 5g
Save £1.20
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15484979
SDS View Documents
Thermo Scientific Alfa Aesar
J62652.14
25g
ONLINE EXCLUSIVE PRICE
£154.00
£151.00 / 25g
Save £3.00
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in acetic acid. Also soluble in acetone or cloroform. Slightly soluble in water

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Chemical Identifiers

70458-96-7
319.336
OGJPXUAPXNRGGI-UHFFFAOYSA-N
4539
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
C16H18FN3O3
MFCD00079532
Baccidal; Sebercim
CHEBI:100246
CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O

Specifications

70458-96-7
C16H18FN3O3
14,6700
Soluble in acetic acid; Also soluble in acetone or cloroform; Slightly soluble in water
CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O
319.336
CHEBI:100246
Norfloxacin
White to Yellow
MFCD00079532
Baccidal; Sebercim
OGJPXUAPXNRGGI-UHFFFAOYSA-N
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
4539
319.33
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 274-614-4

missing translation for 'rtecsNumber' : VB5005000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.