missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin, Thermo Scientific™

A synthetic fluoroquinolone antibiotic

Brand:  Thermo Scientific Alfa Aesar J62652.14

Quantity 25g

 View more versions of this product

Product Code. 15484979

  • ONLINE EXCLUSIVE PRICE
    £154.00
    £151.00 / 25g
    Save £3.00
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in acetic acid. Also soluble in acetone or cloroform. Slightly soluble in water

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

70458-96-7
319.336
OGJPXUAPXNRGGI-UHFFFAOYSA-N
4539
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
C16H18FN3O3
MFCD00079532
Baccidal; Sebercim
CHEBI:100246
CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O

Specifications

70458-96-7
25g
MFCD00079532
Baccidal; Sebercim
OGJPXUAPXNRGGI-UHFFFAOYSA-N
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
4539
319.33
White to Yellow
C16H18FN3O3
14,6700
Soluble in acetic acid; Also soluble in acetone or cloroform; Slightly soluble in water
CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O
319.336
CHEBI:100246
Norfloxacin
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.