missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP10 Antibody (SR1053), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifica
| Antigen | NOP10 |
|---|---|
| Clone | SR1053 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
18626454
|
Novus Biologicals
NBP3-22490-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600164
|
Novus Biologicals
NBP3-22490-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOP10 Monoclonal antibody specifically detects NOP10 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpécification
| NOP10 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| H/ACA ribonucleoprotein complex subunit 3, homolog of yeast Nop10p, MGC70651, NOLA3Nucleolar protein family A member 3, NOP10 ribonucleoprotein homolog (yeast), NOP10P, Nucleolar protein 10, nucleolar protein family A, member 3 (H/ACA small nucleolar RNPs), snoRNP protein NOP10 | |
| A synthesized peptide derived from human NOP10 (Uniprot #: Q9NPE3) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1053 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 55505 | |
| IgG | |
| Affinity purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit