missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP10 Antibody (SR1053), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22490-100ul
This item is not returnable.
View return policy
Description
NOP10 Monoclonal antibody specifically detects NOP10 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| NOP10 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1053 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| H/ACA ribonucleoprotein complex subunit 3, homolog of yeast Nop10p, MGC70651, NOLA3Nucleolar protein family A member 3, NOP10 ribonucleoprotein homolog (yeast), NOP10P, Nucleolar protein 10, nucleolar protein family A, member 3 (H/ACA small nucleolar RNPs), snoRNP protein NOP10 | |
| A synthesized peptide derived from human NOP10 (Uniprot #: Q9NPE3) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55505 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction