Learn More
Brand™ Non-Sterile Polyethylene Filter Tips
Non-self-sealing filter tips from Brand™ have a PE filter that is free from chemical additives.
£49.05 - £105.00
Specifications
Sterility | Non-sterile |
---|---|
For Use With (Application) | Ideal for biological samples that contain detergents such as TRITON™ X-100, SDS, Tween |
Tip Style | Filter Tip |
Graduated | Yes |
Product Code | Brand | Volume (Metric) | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Packaging | Price | Quantity & Availability | ||||
15184957
|
Brand™
732614 |
1000 μL | Racked: 10 TipRacks, 96 each |
£105.00
Pack of 960 |
In Stock
Log in to see stock. |
||||
15154967
|
Brand™
732704 |
0.5 to 10 μL | TipBox: 5 boxes, 96 each |
£54.50
Pack of 480 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
15154957
|
Brand™
732612 |
5 to 200 μL | Racked: 10 TipRacks, 96 each |
£84.30
Pack of 960 |
In Stock
Log in to see stock. |
||||
15114977
|
Brand™
732710 |
5 to 100 μL | TipBox: 5 boxes, 96 each |
£50.35
Pack of 480 |
In Stock
Log in to see stock. |
||||
15114957
|
Brand™
732604 |
0.5 to 10 μL | Racked: 10 tip racks, 96 each |
£95.40
Pack of 960 |
In Stock
Log in to see stock. |
||||
15184967
|
Brand™
732714 |
50 to 1000 μL | TipBox: 5 boxes, 96 each |
£55.70
Pack of 480 |
In Stock
Log in to see stock. |
||||
15164967
|
Brand™
732706 |
1 to 20 μL | TipBox: 5 boxes, 96 each |
£49.05
Pack of 480 |
In Stock
Log in to see stock. |
||||
15194967
|
Brand™
732712 |
4 to 200 μL | TipBox: 5 boxes, 96 each |
£50.35
Pack of 480 |
In Stock
Log in to see stock. |
||||
15174967
|
Brand™
732708 |
2 to 20 μL | TipBox, 5 boxes of 96 each |
£49.05
Pack of 480 |
In Stock
Log in to see stock. |
||||
15194947
|
Brand™
732602 |
0.1 to 1.0 μL | Racked: 10 TipRacks, 96 each |
£103.00
Pack of 960 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
15134957
|
Brand™
732606 |
1 to 20 μL | Racked: 10 TipRacks, 96 each |
£95.40
Pack of 960 |
In Stock
Log in to see stock. |
||||
15124957
|
Brand™
732608 |
2 to 20 μL | Racked: 10 TipRacks, 96 each |
£76.35
Pack of 960 |
In Stock
Log in to see stock. |
||||
15144967
|
Brand™
732702 |
0.1 to 1.0 μL | TipBox: 5 boxes, 96 each |
£58.25
Pack of 480 |
In Stock
Log in to see stock. |
||||
Description
- Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipet shaft
- Functions with consistent reliability
- Sample can be recovered from the filter by simply actuating the pipette’s blow-out function, or by centrifugation if necessary
- Available in a wide range of capacities
Specifications
Non-sterile | |
Ideal for biological samples that contain detergents such as TRITON™ X-100, SDS, Tween | |
Filter Tip | |
Yes |