missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOLC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NOLC1 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16819143
|
Proteintech
11815-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809143
|
Proteintech
11815-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOPP140 (140 kDa nucleolar phosphoprotein) is implicated to play a role in ribosome assembly and the regulation of rRNA transcription. It has been observed to shuttle between the nucleus and cytoplasm and may also be important to nucleolar structural organization.Specifications
| NOLC1 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| E9Q5C9, P41777, Q14978 | |
| 64896, 70769, 9221 | |
| NOLC1 Fusion Protein Ag2382 | |
| Primary | |
| -20°C | |
| NOLC1 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| KIAA0035, NOLC1, NOPP130, NOPP140, NS5ATP13, Nucleolar 130 kDa protein, Nucleolar phosphoprotein p130 | |
| Nolc1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title