missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOLC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11815-1-AP-150UL
This item is not returnable.
View return policy
Description
NOPP140 (140 kDa nucleolar phosphoprotein) is implicated to play a role in ribosome assembly and the regulation of rRNA transcription. It has been observed to shuttle between the nucleus and cytoplasm and may also be important to nucleolar structural organization.Specifications
| NOLC1 | |
| Polyclonal | |
| Unconjugated | |
| NOLC1 | |
| KIAA0035, NOLC1, NOPP130, NOPP140, NS5ATP13, Nucleolar 130 kDa protein, Nucleolar phosphoprotein p130 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64896, 70769, 9221 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| E9Q5C9, P41777, Q14978 | |
| Nolc1 | |
| NOLC1 Fusion Protein Ag2382 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction