missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NOL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | NOL4 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL, ELISA, Immunohistochemistry-Paraffin : 10 μg/mL |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066817
|
Novus Biologicals™
NBP3-30949-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066906
|
Novus Biologicals™
NBP3-30949-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOL4 Polyclonal antibody specifically detects NOL4 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)Specifications
| NOL4 | |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CT125, HRIHFB2255, NOLPcancer/testis antigen 125, nucleolar localized protein, nucleolar protein 4, Nucleolar-localized protein | |
| Synthetic peptide derived from the human NOL4 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1-5 μg/mL, ELISA, Immunohistochemistry-Paraffin : 10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 8715 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title