missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NOL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30949-100ug
This item is not returnable.
View return policy
Description
NOL4 Polyclonal antibody specifically detects NOL4 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| NOL4 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL, ELISA, Immunohistochemistry-Paraffin : 10 μg/mL | |
| CT125, HRIHFB2255, NOLPcancer/testis antigen 125, nucleolar localized protein, nucleolar protein 4, Nucleolar-localized protein | |
| Synthetic peptide derived from the human NOL4 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 8715 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction