missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | NOL1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18334095
|
Novus Biologicals
NBP3-15751-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18391684
|
Novus Biologicals
NBP3-15751-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOL1 Polyclonal antibody specifically detects NOL1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| NOL1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.1.1, EC 2.1.1.-, MGC149287, MGC149288, NOL1/NOP2/Sun domain family, member 1, NOL1MGC117384, NOP120, NOP2 nucleolar protein homolog (yeast), NOP2/Sun domain family, member 1, NSUN1, Nucleolar protein 1, nucleolar protein 1 (120kD), nucleolar protein 1, 120kDa, Nucleolar protein 2 homolog, nucleolar protein 2 homolog (yeast), p120, Proliferating-cell nucleolar antigen p120, Proliferation-associated nucleolar protein p120, putative ribosomal RNA methyltransferase NOP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 355-590 of human NOL1 (NP_001245237.1). | |
| Primary | |
| Store at 4°C. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH7.3 | |
| 4839 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title