missing translation for 'onlineSavingsMsg'
Learn More

NOL1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18391684 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391684 100 μg 100µL
18334095 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18391684 Supplier Novus Biologicals Supplier No. NBP315751100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOL1 Polyclonal antibody specifically detects NOL1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, pH7.3
Gene Alias EC 2.1.1, EC 2.1.1.-, MGC149287, MGC149288, NOL1/NOP2/Sun domain family, member 1, NOL1MGC117384, NOP120, NOP2 nucleolar protein homolog (yeast), NOP2/Sun domain family, member 1, NSUN1, Nucleolar protein 1, nucleolar protein 1 (120kD), nucleolar protein 1, 120kDa, Nucleolar protein 2 homolog, nucleolar protein 2 homolog (yeast), p120, Proliferating-cell nucleolar antigen p120, Proliferation-associated nucleolar protein p120, putative ribosomal RNA methyltransferase NOP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 355-590 of human NOL1 (NP_001245237.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4839
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.