missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Noise-Insulated Vacuum Pump Module With Built-In Pump, For Vo101
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
VO101
Tamaño de la unidad:
Each
Descripción
- Press the main switch on the pump module. This disconnects the appliance from the power supply at all poles.
- Connect the vacuum pump's 230 V plug with earthing contact to the internal socket in the pump module
- Make sure to establish a safe PE conductor connection. Place the power cable so that it is easily accessible at all times and can be pulled out quickly, for example in case of a malfunction or emergency; it does not represent a trip hazard; it cannot come into contact with any hot parts.
Especificaciones
Especificaciones
| Amperage | 1.9 A |
| Depth (Metric) | 550 mm |
| Wattage | 140 W |
| Product Line | Vacuum Pump Module |
| Overall Height (Metric) | 760 mm |
| Overall Width (Metric) | 710 mm |
| For Use With (Equipment) | VO101 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
