missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nogo Antibody (NG-101), Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | Nogo |
|---|---|
| Clone | NG-101 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045183
|
Novus Biologicals™
NBP3-28324-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047952
|
Novus Biologicals™
NBP3-28324-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046409
|
Novus Biologicals™
NBP3-28324-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nogo Monoclonal antibody specifically detects Nogo in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Nogo | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Apoptosis | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| ASY, foocen, Human NogoA, KIAA0886, My043 protein, Nbla00271, Nbla10545, neurite growth inhibitor 220, Neurite outgrowth inhibitor, Neuroendocrine-specific protein, Neuroendocrine-specific protein C homolog, NI220/250, Nogo protein, NOGO-A, Nogo-B, Nogo-C, NOGOC, NOGORTN4-B1, NSP, NSP-CL, reticulon 4, reticulon 5, reticulon-4, reticulon-5, RTN4-A, RTN4-B2, RTN4-C, RTN-x | |
| RTN4/ NOGO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| NG-101 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q9NQC3 | |
| 57142 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title