missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nogo Antibody (NG-101), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28324-1mg
This item is not returnable.
View return policy
Description
Nogo Monoclonal antibody specifically detects Nogo in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Nogo | |
| Monoclonal | |
| Unconjugated | |
| Q9NQC3 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| NG-101 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| ASY, foocen, Human NogoA, KIAA0886, My043 protein, Nbla00271, Nbla10545, neurite growth inhibitor 220, Neurite outgrowth inhibitor, Neuroendocrine-specific protein, Neuroendocrine-specific protein C homolog, NI220/250, Nogo protein, NOGO-A, Nogo-B, Nogo-C, NOGOC, NOGORTN4-B1, NSP, NSP-CL, reticulon 4, reticulon 5, reticulon-4, reticulon-5, RTN4-A, RTN4-B2, RTN4-C, RTN-x | |
| RTN4/ NOGO | |
| 1 mg | |
| Apoptosis | |
| 57142 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction