missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55404
This item is not returnable.
View return policy
Description
NOB1 Polyclonal specifically detects NOB1 in Human samples. It is validated for Western Blot.
Specifications
| NOB1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| adenocarcinoma antigen recognized by T lymphocytes 4, ART4, ART-4, MST158, nin one binding protein, NIN1/RPN12 binding protein 1 homolog (S. cerevisiae), NOB1PPSMD8BP1, Phosphorylation regulatory protein HP-10, Protein ART-4, PSMD8 binding protein 1, RNA-binding protein NOB1 | |
| Rabbit | |
| 47 kDa | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 28987 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9ULX3 | |
| NOB1 | |
| Synthetic peptides corresponding to NOB1(NIN1/RPN12 binding protein 1 homolog (S. cerevisiae)) The peptide sequence was selected from the N terminal of NOB1. Peptide sequence TEIRDKATRRRLAVLPYELRFKEPLPEYVRLVTEFSKKTGDYPSLSATDI. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Yeast: 77%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction