missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00
Specifications
| Antigen | NLRP11 |
|---|---|
| Dilution | Western Blot 0.4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NLRP11 Polyclonal specifically detects NLRP11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NLRP11 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 204801 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HLGNDGVAKLLESLISPDCVLKVVGLPLTGLNTQTQQLLMTVKERKPSLIFLSETWSLKEGREIGVTPASQPGSIIPNSNLDYMFFKFPRMSAAMRTSNTASR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CLR19.6, NACHT, leucine rich repeat and PYD containing 11, NALP11NACHT, LRR and PYD domains-containing protein 11, NLR family, pyrin domain containing 11, NOD17PYPAF7, Nucleotide-binding oligomerization domain protein 17, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 11, PAAD- and NACHT-containing protein 10, PAAD- and NACHT-containing protein 10B, PAAD-and NACHT domain-containing protein 10, PAN10FLJ26273, PYPAF6NACHT, LRR and PYD containing protein 11, PYRIN-containing APAF1-like protein 6 | |
| NLRP11 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title