missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLGN4X Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NLGN4X |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18059335
|
Proteintech
27278-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18019435
|
Proteintech
27278-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NLGN4X antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NLGN4X. This antibody recognizes human, mouse, rat, pig antigen. The NLGN4X antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NLGN4X | |
| Western Blot, ELISA | |
| Rabbit | |
| 57502 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 600 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Pig | |
| ASPGX2, AUTSX2, HLNX, HNLX, KIAA1260, neuroligin 4, X linked, Neuroligin X, NLGN, NLGN4, NLGN4X | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title