Learn More
NLGN4X Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27278-1-AP-20UL
Description
The NLGN4X antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NLGN4X. This antibody recognizes human, mouse, rat, pig antigen. The NLGN4X antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NLGN4X | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ASPGX2, AUTSX2, HLNX, HNLX, KIAA1260, neuroligin 4, X linked, Neuroligin X, NLGN, NLGN4, NLGN4X | |
| 20 μL | |
| 57502 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |