missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.2 Antibody (NX2/1422R), Novus Biologicals™
Rabbit Monoclonal Antibody
£270.00 - £432.00
Specifications
| Antigen | NKX2.2 |
|---|---|
| Clone | NX2/1422R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18250710
|
Novus Biologicals
NBP2-53208-20UG |
20 μg |
£270.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264390
|
Novus Biologicals
NBP2-53208-100UG |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKX2.2 Monoclonal specifically detects NKX2.2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NKX2.2 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B | |
| NKX2-2 | |
| IgG κ | |
| Protein A purified | |
| 45 kDa |
| NX2/1422R | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 4821 | |
| Recombinant human NKX2.2 protein fragment (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title