missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.2 Antibody (NX2/1422R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-53208-20UG
This item is not returnable.
View return policy
Description
NKX2.2 Monoclonal specifically detects NKX2.2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NKX2.2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B | |
| Rabbit | |
| 45 kDa | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| NX2/1422R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| NKX2-2 | |
| Recombinant human NKX2.2 protein fragment (exact sequence is proprietary) | |
| Protein A purified | |
| RUO | |
| 4821 | |
| Store at 4C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction