missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp30/NCR3 Antibody (MM0490-9D31) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £426.00
Specifications
| Antigen | NKp30/NCR3 |
|---|---|
| Clone | MM0490-9D31 |
| Concentration | LYOPH |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006387
|
Novus Biologicals
NBP2-11819-0.025MG |
0.025 mg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18064447
|
Novus Biologicals
NBP2-11819 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| NKp30/NCR3 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q05D23 | |
| 259197 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0490-9D31 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Activating natural killer receptor p30, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NKP30, NKp30MALS | |
| NCR3 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title