missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp30/NCR3 Antibody (MM0490-9D31) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11819-0.025MG
This item is not returnable.
View return policy
Description
NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| NKp30/NCR3 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 1:10-1:1000, CyTOF-ready | |
| Q05D23 | |
| NCR3 | |
| Human NCR3 | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| MM0490-9D31 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| Activating natural killer receptor p30, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NKP30, NKp30MALS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 259197 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction