missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1R Antibody (SR1839), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | NK1R |
|---|---|
| Clone | SR1839 |
| Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605684
|
Novus Biologicals
NBP3-22673-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619873
|
Novus Biologicals
NBP3-22673-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NK1R Monoclonal antibody specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| NK1R | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
| A synthesized peptide derived from human NK1R (Uniprot #: P25103) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1839 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6869 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title