missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1R Antibody (SR1839), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22673-100ul
This item is not returnable.
View return policy
Description
NK1R Monoclonal antibody specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| NK1R | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1839 | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
| A synthesized peptide derived from human NK1R (Uniprot #: P25103) | |
| 100 μg | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6869 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction