missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nitrotyrosine Antibody (2A12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30876-100ug
This item is not returnable.
View return policy
Description
Nitrotyrosine Monoclonal antibody specifically detects Nitrotyrosine in Nonspecies specific samples. It is validated for Western Blot
Specifications
| Nitrotyrosine | |
| Monoclonal | |
| Unconjugated | |
| 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, pH 7.5 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Western Blot | |
| 2A12 | |
| Western Blot : 1:1000 | |
| Nitrotyrosine | |
| 3-Nitrotyrosine-KLH | |
| 100 μg | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction