missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nitrotyrosine Antibody (2A12), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Nitrotyrosine |
|---|---|
| Clone | 2A12 |
| Dilution | Western Blot : 1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067079
|
Novus Biologicals™
NBP3-30876-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066854
|
Novus Biologicals™
NBP3-30876-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nitrotyrosine Monoclonal antibody specifically detects Nitrotyrosine in Nonspecies specific samples. It is validated for Western BlotSpecifications
| Nitrotyrosine | |
| Western Blot : 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, pH 7.5 | |
| 3-Nitrotyrosine-KLH | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 2A12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| Nitrotyrosine | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title