missing translation for 'onlineSavingsMsg'
Learn More

Nitrocyclopentane, 99%

Product Code. 11327955
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327955 5 g 5g
11317955 1 g 1g
11337955 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327955 Supplier Thermo Scientific Alfa Aesar Supplier No. L08029.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2562-38-1 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2562-38-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00001362
InChI Key CJSZWOGCKKDSJG-UHFFFAOYSA-N
Synonym cyclopentane, nitro, nitrocyclopentase, ccris 5057, nitro-cyclopentane, acmc-20ak2p, nitrocyclopentane, cjszwogckkdsjg-uhfffaoysa
PubChem CID 75714
IUPAC Name nitrocyclopentane
SMILES [O-][N+](=O)C1CCCC1

Specifications

Density 1.082
Boiling Point 179°C to 180°C
Flash Point 67°C (152°F)
Refractive Index 1.454
Quantity 5 g
Beilstein 907079
Formula Weight 115.13
Percent Purity 99%
Chemical Name or Material Nitrocyclopentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.