accessibility menu, dialog, popup

UserName

CAS RN 2562-38-1

ONO

CAS RN 2562-38-1

IUPAC Name: nitrocyclopentane
Synonyms: nitrocyclopentane
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: CJSZWOGCKKDSJG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1CCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)