missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Nitrite/pH Test Strip Kits

Product Code. 10632321 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10632321 100 Pack Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10632321 Supplier Macherey-Nagel™ Supplier No. 91338
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Allows for the quick and easy determination of nitrite and pH in cooling lubricants. Macherey Nagel™ Quantofix™ Nitrite/pH Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

  • Ready to use
  • Rapid: provides reliable results within one minute
  • Indicates bacterial growth in the cooling lubricant cycle
  • Helps protect workers against carcinogenic nitrosamine
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C

Order Info

Graduations

Specifications

Analytes Nitrite/pH Test Strip
For Use With (Application) Quick and easy determination of Nitrite and pH in cooling lubricants
Graduations 0 to 80mg/L NO2–, 6.0 to 9.6 pH
Quantity 100 Pack
Type Semi Quantitative Test Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.