missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Nitrite Cuvette Tests
Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Officially approved for consent limits. Environmentally friendly due to less chemicals.
£115.00
Specifications
No. of Tests | 25 |
---|---|
Storage Requirements | 15°C to 25°C |
Test Method | Diazotization |
Type | Nitrite Cuvette Test |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
10205084
|
Hach Lange™
LCK342 |
0.6 to 6mg/L NO3-N |
£115.00
0.30kg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Nitrite cuvette test 0.015- 0.6 mg/L NO2-N
- For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
- EN ISO 26777, DIN 38405 D10 certifications
- Precise and reliable measurement values
- Convenient and error-free dosing of the reagents
- Diazotisation method
- Shelf life of 23 months from production date
- Measuring range: 0.015 – 0.6 mg/L NO2-N
- Measuring range (2): 0.05 - 2 mg/L NO2
Specifications
25 | |
15°C to 25°C | |
Diazotization | |
Nitrite Cuvette Test |