missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Nitrite Cuvette Tests
Description
- Nitrite cuvette test 0.015- 0.6 mg/L NO2-N
- For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
- EN ISO 26777, DIN 38405 D10 certifications
- Precise and reliable measurement values
- Convenient and error-free dosing of the reagents
- Diazotisation method
- Shelf life of 23 months from production date
- Measuring range: 0.015 – 0.6 mg/L NO2-N
- Measuring range (2): 0.05 - 2 mg/L NO2
Specifications
Specifications
| No. of Tests | 25 |
| Range | 0.6 to 6mg/L NO3-N |
| Storage Requirements | 15°C to 25°C |
| Test Method | Diazotization |
| Type | Nitrite Cuvette Test |
Safety and Handling
Product Identifier
- Nitrit/Nitrite, DosiCapZip
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- MIXTURE LIST-Contain: 1-Naphthylamine sulfonic acid salt, Sulphanilic acid sodiumsalt, Sodium hydroxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?