missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nitrile Indigo Disposable Gloves PPE Cat III

Product Code. 17102192 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17102192 X-Large Pack of 200
17112192 X-Small Pack of 200
17192182 Small Pack of 200
17182182 Medium Pack of 200
17172182 Large Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17102192

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fisherbrand™ non-sterile non-powdered gloves offer excellent protection for a wide range of applications and general use, while the textured fingertips provide good grip.

  • PPE Category III
  • Indigo blue color
  • Nitrile
  • Ambidextrous
  • Beaded cuff
  • Textured fingertips
  • Non-powdered
  • General purpose use
  • Chemical resistant
  • Disposable
  • Non-sterile

Specifications

Certifications/Compliance PPE Category III, EN 420:2003, EN ISO 374-1/Type C, EN ISO 374-5, EN ISO 374-4
Content And Storage Shelf Life 5 Years
Applications Laboratory
AQL 1.5
Color Indigo
Disposable Single-use
Finger Thickness (Metric) 0.08 to 0.14 mm
Hand Ambidextrous
Material Nitrile
Palm Thickness (Metric) 0.06 to 0.09 mm
Size X-Large
Sterility Non-sterile
Thickness (Metric) 0.05 to 0.08 mm (Cuff)
Gender Unisex
Glove Cuff Style Beaded
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.