missing translation for 'onlineSavingsMsg'
Learn More

Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Take advantage of a quality product that delivers accurate results with the Epredia™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

£131.00 - £691.00

Specifications

Antigen Nitric Oxide Synthase, inducible (iNOS) Ab-1
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Formulation Price Quantity & Availability  
Product Code Brand Quantity Formulation Price Quantity & Availability  
12603077
View Documents
Epredia
RB-1605-P0
100 μL Purified with BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12613077
View Documents
Epredia
RB-1605-P1
500 μL Purified with BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12623077
View Documents
Epredia
RB-1605-P1ABX
500 μL Purified w/o BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12673067
View Documents
Epredia
RB-1605-P
1 mL Purified with BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12633077
View Documents
Epredia
RB-1605-PABX
1 mL Purified w/o BSA and Azide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Rat, Mouse and Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages

Molecular Weight: 130kDa

Postitive Control: Rat lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Nitric Oxide Synthase, inducible (iNOS) Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Neuroscience
A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages
Primary
1mg/mL
Polyclonal
Concentrated
RUO
Human, Mouse
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.