Learn More
Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Take advantage of a quality product that delivers accurate results with the Epredia™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.
Brand: Epredia RB-1605-P0
Description
NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Rat, Mouse and Human. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages
Molecular Weight: 130kDa
Postitive Control: Rat lung
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
| Nitric Oxide Synthase, inducible (iNOS) Ab-1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| 100 μL | |
| Neuroscience | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Purified with BSA and Azide | |
| A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.