missing translation for 'onlineSavingsMsg'
Learn More

Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Take advantage of a quality product that delivers accurate results with the Epredia™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-1605-P

Code : LD

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 12673067

  • £684.00 / 1mL
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Rat, Mouse and Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages

Molecular Weight: 130kDa

Postitive Control: Rat lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Nitric Oxide Synthase, inducible (iNOS) Ab-1
Polyclonal
1mg/mL
Rabbit
1 mL
Neuroscience
Human, Mouse
IgG
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers