Learn More
Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Take advantage of a quality product that delivers accurate results with the Epredia™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.
Brand: Epredia RB-1605-PABX
Code : LD
Additional Details : Weight : 0.00100kg
Description
NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Rat, Mouse and Human. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages
Molecular Weight: 130kDa
Postitive Control: Rat lung
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
Nitric Oxide Synthase, inducible (iNOS) Ab-1 | |
Polyclonal | |
Unconjugated | |
A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages | |
RUO | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
1mg/mL | |
Rabbit | |
1 mL | |
Neuroscience | |
Human, Mouse | |
IgG |