missing translation for 'onlineSavingsMsg'
Learn More

Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Take advantage of a quality product that delivers accurate results with the Epredia™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-1605-P1ABX

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12623077

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Rat, Mouse and Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages

Molecular Weight: 130kDa

Postitive Control: Rat lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Nitric Oxide Synthase, inducible (iNOS) Ab-1
Polyclonal
Unconjugated
A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
1mg/mL
Rabbit
500 μL
Neuroscience
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers