missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92656-0.1ml
This item is not returnable.
View return policy
Description
CLCN5 Polyclonal antibody specifically detects CLCN5 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CLCN5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| chloride channel 5, Chloride channel protein 5, Chloride transporter ClC-5, clC-5, CLC5, CLCK2NPHL2, DENTSNPHL1, H(+)/Cl(-) exchange transporter 5, hCIC-K2, hClC-K2, nephrolithiasis 1 (X-linked), nephrolithiasis 2, X-linked, XLRH, XRN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 637-816 of human CLCN5 (NP_001121370.1). YPFLEAKEEFAHKTLAMDVMKPRRNDPLLTVLTQDSMTVEDVETIISETTYSGFPVVVSRESQRLVGFVLRRDLIISIENARKKQDGVVSTSIIYFTEHSPPLPPYTPPTLKLRNILDLSPFTVTDLTPMEIVVDIFRKLGLRQCLVTHNGRLLGIITKKDVLKHIAQMANQDPDSILFN | |
| 0.1 mL | |
| DNA Repair, Signal Transduction | |
| 1184 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction