missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | CLCN5 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617952
|
Novus Biologicals
NBP2-92656-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686962
|
Novus Biologicals
NBP2-92656-0.1ml |
0.1 mL |
£359.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLCN5 Polyclonal antibody specifically detects CLCN5 in Human, Mouse samples. It is validated for Western BlotSpecifications
| CLCN5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1184 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| chloride channel 5, Chloride channel protein 5, Chloride transporter ClC-5, clC-5, CLC5, CLCK2NPHL2, DENTSNPHL1, H(+)/Cl(-) exchange transporter 5, hCIC-K2, hClC-K2, nephrolithiasis 1 (X-linked), nephrolithiasis 2, X-linked, XLRH, XRN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 637-816 of human CLCN5 (NP_001121370.1). YPFLEAKEEFAHKTLAMDVMKPRRNDPLLTVLTQDSMTVEDVETIISETTYSGFPVVVSRESQRLVGFVLRRDLIISIENARKKQDGVVSTSIIYFTEHSPPLPPYTPPTLKLRNILDLSPFTVTDLTPMEIVVDIFRKLGLRQCLVTHNGRLLGIITKKDVLKHIAQMANQDPDSILFN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title