missing translation for 'onlineSavingsMsg'
Learn More

NIRF Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93806-0.1ml

 View more versions of this product

Product Code. 18678080

  • £366.00 / 0.01mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NIRF Polyclonal antibody specifically detects NIRF in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NIRF
Polyclonal
Western Blot 1:500 - 1:1000
DKFZp434B0920, DKFZp686G0837, E3 ubiquitin-protein ligase UHRF2, EC 6.3.2, EC 6.3.2.-, MGC33463, NIRFUbiquitin-like-containing PHD and RING finger domains protein 2, Np95/ICBP90-like RING finger protein, Np95-like RING finger protein, Nuclear protein 97, Nuclear zinc finger protein Np97, RNF107RING finger protein 107, Ubiquitin-like PHD and RING finger domain-containing protein 2, ubiquitin-like with PHD and ring finger domains 2, ubiquitin-like, containing PHD and RING finger domains, 2, URF2
Recombinant fusion protein containing a sequence corresponding to amino acids 76-359 of mouse NIRF (NP_659122.2). DSSLPSTSKQNDAQVKPSSHNPPKVKKTARGGSSSQPSTSARTCLIDPGFGLYKVNELVDARDVGLGAWFEAHIHSVTRASDGHSRGKTPLKNGSSYKRTNGNVNHNSKENTNKLDNVPSTSNSDSVAADEDVIYHIEYDEYPESGILEMNVKDLRPRARTILKWNELNVGDVVMVNYNVENPGKRGFWYDAEITTLKTISRTKKEVRVKVFLGGSEGTLNDCRVMSVDEIFKIEKPGAHPISFADGKFLRKNDPECDLCGGDPDKTCHMCSCHKCGEKRDPNM
0.1 mL
Cancer, Zinc Finger
115426
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.