missing translation for 'onlineSavingsMsg'
Learn More

NIRF Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£164.00 - £366.00

Specifications

Antigen NIRF
Dilution Western Blot 1:500 - 1:1000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18697040
View Documents
Novus Biologicals
NBP2-93806-0.02ml
0.02 mL
£164.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18678080
View Documents
Novus Biologicals
NBP2-93806-0.1ml
0.1 mL
£366.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NIRF Polyclonal antibody specifically detects NIRF in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

NIRF
Western Blot
Unconjugated
Rabbit
Cancer, Zinc Finger
PBS (pH 7.3), 50% glycerol
115426
IgG
Affinity purified
Western Blot 1:500 - 1:1000
Polyclonal
Purified
RUO
Human, Mouse, Rat
DKFZp434B0920, DKFZp686G0837, E3 ubiquitin-protein ligase UHRF2, EC 6.3.2, EC 6.3.2.-, MGC33463, NIRFUbiquitin-like-containing PHD and RING finger domains protein 2, Np95/ICBP90-like RING finger protein, Np95-like RING finger protein, Nuclear protein 97, Nuclear zinc finger protein Np97, RNF107RING finger protein 107, Ubiquitin-like PHD and RING finger domain-containing protein 2, ubiquitin-like with PHD and ring finger domains 2, ubiquitin-like, containing PHD and RING finger domains, 2, URF2
Recombinant fusion protein containing a sequence corresponding to amino acids 76-359 of mouse NIRF (NP_659122.2). DSSLPSTSKQNDAQVKPSSHNPPKVKKTARGGSSSQPSTSARTCLIDPGFGLYKVNELVDARDVGLGAWFEAHIHSVTRASDGHSRGKTPLKNGSSYKRTNGNVNHNSKENTNKLDNVPSTSNSDSVAADEDVIYHIEYDEYPESGILEMNVKDLRPRARTILKWNELNVGDVVMVNYNVENPGKRGFWYDAEITTLKTISRTKKEVRVKVFLGGSEGTLNDCRVMSVDEIFKIEKPGAHPISFADGKFLRKNDPECDLCGGDPDKTCHMCSCHKCGEKRDPNM
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.