missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIP30 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NIP30 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16896584
|
Proteintech
16830-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16886584
|
Proteintech
16830-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM192A is also known as Nip30. This protein has not bee characterized.Specifications
| NIP30 | |
| Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q91WE2, Q9GZU8 | |
| 102122, 307652, 80011 | |
| NIP30 Fusion Protein Ag10439 | |
| Primary | |
| -20°C | |
| FAM192A |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C16orf94, CDA018, CDA10, FAM192A, NIP30, Protein FAM192A | |
| Fam192a | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title