missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIP30 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16830-1-AP-150UL
This item is not returnable.
View return policy
Description
FAM192A is also known as Nip30. This protein has not bee characterized.Specifications
| NIP30 | |
| Polyclonal | |
| Unconjugated | |
| FAM192A | |
| C16orf94, CDA018, CDA10, FAM192A, NIP30, Protein FAM192A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102122, 307652, 80011 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91WE2, Q9GZU8 | |
| Fam192a | |
| NIP30 Fusion Protein Ag10439 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction